missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4b Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marca: Invitrogen PA170042
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-70042 detects C4b in human samples. PA1-70042 has been successfully used in flow cytometry, Western blot and ELISA procedures. The PA1-70042 immunogen is purified human complement C4b.
This gene encodes the basic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene. In addition, this gene exists as a long form and a short form due to the presence or absence of a 6.4 kb endogenous HERV-K retrovirus in intron 9.
Especificaciones
| Complement C4b | |
| Polyclonal | |
| Unconjugated | |
| C4B | |
| Acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; c4 complement; C4A; C4a anaphylatoxin; C4A13; C4A91; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement C4-B-like; complement component 4B (Chido blood group); complement component 4B (Chido blood group), copy 2; CPAMD2; CPAMD3; DADB-112B14.11; FLJ60561; MGC164979 | |
| Chicken | |
| purified | |
| RUO | |
| 100293534, 721 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P0C0L5 | |
| C4B, C4B_2 | |
| Purified human complement C4b. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido