missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Complement C3a Monoclonal Antibody (K13/16-5.7)
GREENER_CHOICE

Product Code. 11514192
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
400 μL
missing translation for 'unitSize'
200 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
11514192 400 μL 200 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11514192 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' GAU0131602

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAU 013-16-02 detects Complement Componet C3a/C3a (desArg) in human samples. This antibody does not cross-react with C4a or C5a. GAU 013-16-02 has been successfully used in ELISA and Western blot procedures. GAU 013-16-02 can be used as a capture antibody in sandwich ELISA with GAU 017-01-02 as detection antibody. This antibody detects an epitope that is present on human C3, C3a, and C3a (desArg), but does not cross react with C4a or C5a. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Complement C3a
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon K13/16-5.7
Concentración 1.07 mg/mL
Conjugado Unconjugated
Formulación PBS with 15mM sodium azide; pH 7.4
génica C3
N.º de referencia del gen P01024
Alias de gen Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3
Símbolos de los genes C3
Especie del huésped Mouse
Inmunógeno Human C3a.
Método de purificación Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 718
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.