missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL4A4 Polyclonal Antibody
GREENER_CHOICE

Código de producto. p-200125862
Change view
Click to view available options
Cantidad:
20 μL
200 μL
Tamaño de la unidad:
20 microlitros
Código de producto. Cantidad unitSize
17740454 200 μL -
17703484 20 μL 20 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17740454

Marca: Invitrogen™ PA5119041

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

COL4A4 Polyclonal Antibody for Western Blot, ICC/IF

Collagens are highly conserved and are characterized by an uninterrupted Glycine-X-Y triplet repeat. Col1, the fibrillar collagen found in most connective tissues, is the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Col2, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia. Col3 is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, aortic and arterial aneurysms. Col4 is a major consitiuent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Col4 antibodies are useful in detecting the loss of parts of basement membrane in carcinomas. Col5 is found in tissues containing Col1 and appears to regulate the assembly of heterotypic fibers composed of both Col1 and Col5. Col5 is closely related to type XI collagen; mutations in this gene are associated with Ehlers-Danlos syndrome.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno COL4A4
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
génica COL4A4
N.º de referencia del gen P53420
Alias de gen CA44; COL4A4; Collagen 4; Collagen alpha-4(IV) chain; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 4 chain; collagen, type IV, alpha 4
Símbolos de los genes COL4A4
Especie del huésped Rabbit
Inmunógeno Recombinant protein Collagen Type IV Alpha 4. The antigen corresponds to amino acid range 1469-1690 of the target protein.
Método de purificación Antigen affinity chromatography, Protein A
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1286
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.