missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Coilin Polyclonal Antibody
Descripción
Recommended positive controls: HepG2 , HepG2 nuclear extract, 293T nuclear extract. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.
Especificaciones
Especificaciones
| Antígeno | Coilin |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.66 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| génica | COIL |
| N.º de referencia del gen | P38432 |
| Alias de gen | C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11 |
| Símbolos de los genes | COIL |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?