Learn More
Melford Coelenterazine Native
Descripción
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion.
Cell permeable aequorin luminophore that acts as a sensitive and specific chemiluminescence probe of the superoxide anion. Used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.
Especificaciones
Especificaciones
| CAS | 55779-48-1 |
| Fórmula molecular | C26H21N3O3 |
| Cantidad | 1 mg |