Learn More
Invitrogen™ CNTO 607 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Código de producto. 30283578
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
30283578 100 μg 100 microgramos
30284193 1 mg 1 miligramo
2 options
Código de producto. 30283578
Marca: Invitrogen™ MA559169
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term CNTO 607.
IL13 (Interleukin-13) is a gene located on chromosome 5q31, encoding the IL-13 cytokine, which plays a pivotal role in the immune system, particularly in the regulation of inflammatory and immune responses. IL-13 is predominantly secreted by Th2 cells and is involved in the pathogenesis of allergic reactions and chronic inflammatory conditions such as asthma and ulcerative colitis. This cytokine exerts its effects by binding to the IL-13 receptor complex, which includes IL-13R aplha 1 and IL-4R alpha. Upon binding, IL-13 activates several signaling pathways, including the JAK-STAT pathway, leading to the transcription of genes involved in the immune response. IL-13 is also implicated in tissue remodeling and fibrosis due to its ability to stimulate fibroblast proliferation and collagen production. Dysregulation of IL-13 expression or signaling is associated with various diseases, making it a target for therapeutic intervention, particularly in allergic and autoimmune conditions.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CNTO 607 Biosimilar
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1