Learn More
CNN1, anti-Human, Clone: CNN1/1408R, Abnova™
Monoclonal Antibody
Marca:  Abnova RAB00326.100ug
Código de producto. 16052308
  • 559.00€ / 100 microgramos
Especificaciones
CNN1
Monoclonal
Unconjugated
In 10 mM PBS.
SMCC/Sm-Calp
Original antibody is raised against recombinant protein corresponding to full length human CNN1.
RUO
1264
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Liquid
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
CNN1/1408R
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
calponin 1, basic, smooth muscle
CNN1
100 μg
Primary
Human, Rat
Antibody
IgG κ
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones