Learn More
Invitrogen™ CLPH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA528518
Descripción
Recommended positive controls: Molt-4. Predicted reactivity: Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CLPH (Casein-Like phosphoprotein) is a recently discovered, evolutionarily conserved, testis-specific cytoplasmic protein expressed exclusively in postmeiotic germ cells in rat spermatogenesis. CLPH is a novel casein kinase 2-phosphorylated disordered and calcium-binding protein that displays strong variation in relative expression levels during spermatogenesis. It is frequently associated with the mitochondrial inner membrane during the last steps of spermatid differentiation and is involved in the extremely complex structural rearrangements occurring in haploid germ cells during spermiogenesis.
Especificaciones
| CLPH | |
| Polyclonal | |
| Unconjugated | |
| CABS1 | |
| C4orf35; CABS1; calcium binding protein, spermatid associated 1; calcium-binding and spermatid-specific protein 1; calcium-binding protein, spermatid-specific 1; calcium-binding protein, sperm-specific 1; casein-like phosphoprotein; Clph; NYD-SP26; Protein RSD-6; Rsd6; RSD-6; Testis development protein NYD-SP26 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85438 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96KC9 | |
| CABS1 | |
| Recombinant protein encompassing a sequence within the center region of human NYD-SP26. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.