Learn More
ClozapineClozapine N-oxide, MedChemExpress
Código de producto. 16467256
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16467256 100 mg 100 miligramos
16493317 50 mg 50 miligramos
16457308 10 mg 10 miligramos
16419817 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16467256 Proveedor Medchem Express N.º de proveedor HY17366
MedChemExpress Clozapine N-oxide is a major metabolite of Clozapine and a human muscarinic designer receptors (DREADDs) agonist. Clozapine N-oxide activates the DREADD receptor hM3Dq and hM4Di. Clozapine N-oxide can cross the blood-brain barrier. Clozapine is a potent dopamine antagonist and also a potent and selective muscarinic M4 receptor (EC50=11 nM) agonist.
Clozapine N-oxide is a major metabolite of Clozapine and a human muscarinic designer receptors (DREADDs) agonist. Clozapine N-oxide activates the DREADD receptor hM3Dq and hM4Di. Clozapine N-oxide can cross the blood-brain barrier. Clozapine is a potent dopamine antagonist and also a potent and selective muscarinic M4 receptor (EC50=11 nM) agonist.
Especificaciones
Nombre del producto químico o material ClozapineClozapine N-oxide
Forma física Solid
CAS 34233-69-7
Color Yellow
Cantidad 100 mg
Notas de grado de pureza Research
Fórmula molecular C18H19ClN4O
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Información de solubilidad DMSO : ≥ 100 mg/mL (291.70 mM) ∣H2O : 1 mg/mL (2.92 mM; Need ultrasonic)
SMILES [O-][N+]1(C)CCN(C2=NC3=CC(Cl)=CC=C3NC4=CC=CC=C42)CC1
Peso molecular (g/mol) 342.82
Formula Weight (peso de la fórmula) 342.82
Porcentaje de pureza 99.25%