missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloxacillin sodium monohydrate, MedChemExpress
Description
Cloxacillin sodium monohydrate is an orally active antibacterial agent and β-lactamase inhibitor with an IC50 of 0.04 μM. Cloxacillin sodium monohydrate can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB and NLRP3-related proteins.
Specifications
Specifications
| Nombre del producto químico o material | Cloxacillin sodium monohydrate |
| Forma física | Solid |
| CAS | 7081-44-9 |
| Color | Blanco |
| Cantidad | 100 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C19H19ClN3NaO6S |
| Para utilizar con (aplicación) | COVID-19-immunoregulation |
| Información de solubilidad | DMSO : 100 mg/mL (210.14 mM; Need ultrasonic) ∣H2O : 50 mg/mL (105.07 mM; Need ultrasonic) |
| SMILES | O=C(C(C(C(C=CC=C1)=C1Cl)=NO2)=C2C)N[C@H]3[C@](SC(C)(C)[C@@H]4C(O[Na])=O)([H])N4C3=O.O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?