Learn More
STEMCELL Technologies ClonaCell™-HY Liquid HAT Selection Medium
Código de producto. 17148211
Encompass
Change view
Click to view available options
Cantidad:
500 mL
Tamaño de la unidad:
500 ml
17148211 500 mL 500 ml
1 options
Código de producto. 17148211
Marca: STEMCELL Technologies 03831
Hybridoma selection medium with HAT (serum-containing)
  • ClonaCell™-HY Liquid HAT Selection Medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells
  • This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster
Especificaciones
Tipo de célula Hibridomas
Para utilizar con (aplicación) Cell Culture; Hybridoma Generation
Cantidad 500 mL