Learn More
Clemastine fumarate, MedChemExpress
Código de producto. 16412096
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 mg
200 mg
500 mg
Tamaño de la unidad:
100 miligramos
200 miligramos
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16412096 500 mg 500 miligramos
16471406 200 mg 200 miligramos
16438226 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16412096 Proveedor Medchem Express N.º de proveedor HYB0298A
MedChemExpress Clemastine (HS-592) fumarate is a selective histamine H1 receptor antagonist. Clemastine fumarate is an antihistamine mainly used for relieving symptoms of allergic reactions primarily by competing with histamine to bind H1 receptors. Anti-inflammatory effects.
Clemastine (HS-592) fumarate is a selective histamine H1 receptor antagonist. Clemastine fumarate is an antihistamine mainly used for relieving symptoms of allergic reactions primarily by competing with histamine to bind H1 receptors. Anti-inflammatory effects.
Especificaciones
Nombre del producto químico o material Clemastine fumarate
CAS 14976-57-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C25H30ClNO5
Cantidad 500 mg
Sinónimo HS-592 fumarateMeclastine fumarate
Información de solubilidad DMSO : 14.29 mg/mL (31.07 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.46 mM; Need ultrasonic)
SMILES CN1[C@@H](CCO[C@](C2=CC=CC=C2)(C3=CC=C(Cl)C=C3)C)CCC1.O=C(O)/C=C/C(O)=O
Peso molecular (g/mol) 459.96
Formula Weight (peso de la fórmula) 459.96
Porcentaje de pureza 99.41%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation