missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC4A Monoclonal Antibody (320511)

Descripción
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) DCIR or recombinant mouse DCAR is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in inflammatory and immune response. Multiple transcript variants encoding distinct isoforms have been identified for this gene. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Especificaciones
Especificaciones
| Antígeno | CLEC4A |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 320511 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5% trehalose and No Preservative |
| génica | Clec4a2 |
| N.º de referencia del gen | Q9QZ15 |
| Alias de gen | CD367; CLEC4A; Clec4a2; Clecsf6; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 6; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6; C-type lectin DDB27; C-type lectin domain family 4 member A; C-type lectin domain family 4, member A; C-type lectin domain family 4, member a2; C-type lectin superfamily member 6; DCIR; Dcir1; Dcir2; DDB27; Dendritic cell immunoreceptor; dendritic cell inhibitory receptor 1; dendritic cell inhibitory receptor 2; HDCGC13P; immunoreceptor; lectin-like immunoreceptor; LLIR |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?