Learn More
Cole-Parmer Cleanroom Port Disk Hose Barb Fittings, Flat Plate, Natural Kynar™ Flex
Código de producto. 30053653 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/2 in
1/4 in
1/8 in
1 in
3/16 in
3/4 in
3/8 in
5/16 in
5/32 pulg
5/8 in
Tamaño de la unidad:
paquete de 10
Código de producto. Diámetro interior (imperial) unitSize
30053653 5/16 in paquete de 10
30053717 1 in paquete de 10
30054341 3/4 in paquete de 10
30054265 3/8 in paquete de 10
30053272 1/4 in paquete de 10
30053350 3/16 in paquete de 10
30053176 5/8 in paquete de 10
30053102 1/2 in paquete de 10
30053586 5/32 pulg paquete de 10
30054380 1/8 in paquete de 10
10 options
Código de producto. 30053653
Marca: Cole-Parmer 5011719
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Flexible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to Kynar bags. Meets FDA requirements 21 CFR 177.1520(c), 21 CFR 176.170(c), 21 CFR 177.2600(c)(4)(i), NSF/ANSI Standard 51 and 61, and RoHS compliant
Especificaciones
Certificaciones/conformidad FDA;Non-DEHP;Non-Phthalate;RoHS
Diámetro interior (imperial) 5/16 in
Cantidad de embalaje 10/Pk.
Puerto de salida Port disk x Hose barb
Método de esterilización 121°C
Forma Straight
Color Natural