Learn More
Cole-Parmer Cleanroom Port Disk Hose Barb Fittings, Flat Plate, Engage™ 8402
Tienda Cole-Parmer ProductosDescripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Fle in., ible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to polyethylene bags. Meets FDA requirements 21 CFR 177.1520(c) 3.2c and 21 CFR 176.170(c) and RoHS compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diámetro interior (imperial) | 3/4 in |
| Cantidad de embalaje | 10/Pk. |
| Puerto de salida | Port Disk x Hose Barb |
| Método de esterilización | Gamma irradiation up to 50 kGy |
| Forma | Straight |
| Color | Translúcido |