Learn More
Cole-Parmer Cleanroom Male Luer Plugs, Straight Adapter
Código de producto. 30054378 Tienda Cole-Parmer Productos
Change view
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nailon
Polipropileno
Color:
Blanco
Natural
Transparente
Tamaño de la unidad:
paquete de 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Material Color unitSize
30054378 Kynar Natural paquete de 10
30054116 Nailon Blanco paquete de 10
30053090 Polipropileno Transparente paquete de 10
30053329 Nailon Natural paquete de 10
30053396 CrystalVu (CVU) Transparente paquete de 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 30054378 Proveedor Cole-Parmer N.º de proveedor 5011034
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Especificaciones
Certificaciones/conformidad Non-DEHP;Non-Phthalate;USP
Material Kynar
Conexión Male Luer, Plug
Color Natural