Learn More
Cole-Parmer Cleanroom Male Luer to Hose Barb Fittings, Straight Adapter, Natural Nylon
Código de producto. p-200105178 Tienda Cole-Parmer Productos
Click to view available options
Diámetro interior (imperial):
1/16 in
1/4 in
1/8 in
3/16 in
3/32 in
5/32 pulg
Tamaño de la unidad:
paquete de 10
Código de producto. 30053731
Marca: Cole-Parmer 5011005
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. RoHS compliant
Especificaciones
Certificaciones/conformidad Non-DEHP;Non-Phthalate;RoHS
Diámetro interior (imperial) 3/32 in
Material Nailon
Cantidad de embalaje 10/Pk.
Conexión Male Luer, Hose barb
Color Natural