Learn More
Cole-Parmer Cleanroom Male Luer to Hose Barb Fittings, Straight Adapter, Natural Kynar™
Tienda Cole-Parmer ProductosDescripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;USP |
| Diámetro interior (imperial) | 3/16 in |
| Material | Kynar |
| Cantidad de embalaje | 10/Pk. |
| Conexión | Male Luer, Hose barb |
| Color | Natural |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.