Learn More
Cole-Parmer Cleanroom Large-Bore Male Luer Plugs, Straight Adapter
Código de producto. 30054358 Tienda Cole-Parmer Productos
Change view
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nailon
Polipropileno
Tamaño de la unidad:
paquete de 10
Color:
Blanco
Natural
Transparente
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Material unitSize Color
30054358 Polipropileno paquete de 10 -
30053408 Kynar paquete de 10 Natural
30053859 Nailon paquete de 10 Blanco
30052982 CrystalVu (CVU) paquete de 10 Transparente
30054182 Nailon paquete de 10 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 30054358 Proveedor Cole-Parmer N.º de proveedor 5010950
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Material Polipropileno
Conexión Large Bore Male Luer, Plug
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.