Learn More
Cole-Parmer Cleanroom Large-Bore Female Luer to Hose Barb Fittings, Straight Adapter, White Nylon
Tienda Cole-Parmer ProductosDescripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Large-bore luer fittings offer higher flow rates compared to standard luers. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diámetro interior (imperial) | 5/16 in |
| Material | Nailon |
| Cantidad de embalaje | 10/Pk. |
| Conexión | Large Bore Female Luer, Hose Barb |
| Color | Blanco |