Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Tee Union, White Nylon
Código de producto. p-200105113 Tienda Cole-Parmer Productos
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/8 in
3/16 in
3/18 in
3/32 in
5/32 pulg
Tamaño de la unidad:
paquete de 10
Código de producto. 30053352
Marca: Cole-Parmer 5011087
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Especificaciones
Certificaciones/conformidad FDA;Non-DEHP;Non-Phthalate;RoHS
Diámetro interior (imperial) 1/16 in
Cantidad de embalaje 10/Pk.
Puerto de salida Hose Barb Tee
Método de esterilización E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Forma Tee
Color Blanco