Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diámetro interior (imperial) | 5/32 pulg |
| Cantidad de embalaje | 10/Pk. |
| Puerto de salida | Hose Barb Tee |
| Método de esterilización | E-beam/gamma irradiation, EtO, or autoclave up to 121°C |
| Forma | Tee |
| Color | Blanco |