Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | USP;Non-DEHP;Non-Phthalate |
| Diámetro interior (imperial) | 5/32 pulg |
| Temperatura mín. de funcionamiento (métrico) | -35 °C |
| Cantidad de embalaje | 10/Pk. |
| Puerto de salida | Hose Barb Tee |
| Método de esterilización | E-beam/gamma irradiation, EtO, or autoclave up to 135°C |
| Forma | Tee |
| Color | Natural |