Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, White Nylon
Código de producto. 30054220 Tienda Cole-Parmer Productos
Change view
Click to view available options
Tamaño de los tubos:
De 1/4 a 1/8 in
De 1/4 a 3/16 in
De 1/4 a 5/32 in
De 1/8 a 1/16 in
De 1/8 a 3/32 in
De 3/16 a 1/8 in
De 3/16 a 3/32 in
De 3/32 a 1/16 in
De 3/8 a 1/4 in
De 5/16 a 1/4 in
De 5/32 a 1/8 in
De 5/32 a 3/32 in
Tamaño de la unidad:
paquete de 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño de los tubos unitSize
30054220 De 5/32 a 3/32 in paquete de 10
30053541 De 5/16 a 1/4 in paquete de 10
30054315 De 1/8 a 3/32 in paquete de 10
30054232 De 1/4 a 1/8 in paquete de 10
30053832 De 1/8 a 1/16 in paquete de 10
30052998 De 3/8 a 1/4 in paquete de 10
30053331 De 3/32 a 1/16 in paquete de 10
30054097 De 1/4 a 3/16 in paquete de 10
30054347 De 5/32 a 1/8 in paquete de 10
30053490 De 3/16 a 3/32 in paquete de 10
30053885 De 3/16 a 1/8 in paquete de 10
30054161 De 1/4 a 5/32 in paquete de 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Código de producto. 30054220 Proveedor Cole-Parmer N.º de proveedor 5010868
Solo quedan null
Añadir a la cesta
Edge
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Especificaciones
Certificaciones/conformidad FDA;Non-DEHP;Non-Phthalate;RoHS
Cantidad de embalaje 10/Pk.
Tamaño de los tubos De 5/32 a 3/32 in
Puerto de salida Barb x Barb
Método de esterilización E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Forma Straight
Color Blanco
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.