Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Natural Kynar™
Código de producto. 30053878 Tienda Cole-Parmer Productos
Change view
Click to view available options
Tamaño de los tubos:
De 1 a 1/2 in
De 1 a 3/4 in
De 1 a 5/8 in
De 1/2 a 1/4 in
De 1/2 a 3/8 in
De 1/2 a 5/16 in.
De 1/4 a 1/16 in
De 1/4 a 1/8 in
De 1/4 a 3/16 in
De 1/4 a 3/32 in
De 1/4 a 5/32 in
De 1/8 a 1/16 in
De 1/8 a 3/32 in
De 3/16 a 1/16 in
De 3/16 a 1/8 in
De 3/32 a 1/16 in
De 3/4 a 1/2 in
De 3/4 a 3/8 in
De 3/8 a 1/4 in
De 3/8 a 1/8 in
De 3/8 a 5/16 in
De 3/8 a 5/32 in
De 5/16 a 1/4 in
De 5/16 a 3/16 in
De 5/16 a 5/32 in
De 5/32 a 1/8 in
De 5/32 a 3/32 in
De 5/8 a 1/2 in
De 5/8 a 1/4 in
De 5/8 a 3/8 in
De 5/8 a 5/16 in
Tamaño de la unidad:
paquete de 10
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño de los tubos unitSize
30053878 De 5/32 a 3/32 in paquete de 10
30054257 De 1/8 a 1/16 in paquete de 10
30053205 De 5/8 a 3/8 in paquete de 10
30053467 De 1/2 a 1/4 in paquete de 10
30054351 De 3/8 a 5/16 in paquete de 10
30053539 De 1 a 1/2 in paquete de 10
30053462 De 5/16 a 1/4 in paquete de 10
30053074 De 5/8 a 5/16 in paquete de 10
30053589 De 3/32 a 1/16 in paquete de 10
30054283 De 1/2 a 5/16 in. paquete de 10
30052967 De 5/8 a 1/4 in paquete de 10
30053511 De 1/2 a 3/8 in paquete de 10
30054195 De 3/8 a 1/4 in paquete de 10
30053347 De 1/4 a 1/8 in paquete de 10
30054168 De 3/4 a 3/8 in paquete de 10
30053679 De 5/8 a 1/2 in paquete de 10
30053985 De 3/8 a 5/32 in paquete de 10
30053150 De 5/32 a 1/8 in paquete de 10
30053937 De 1 a 3/4 in paquete de 10
30054377 De 1/4 a 1/16 in paquete de 10
30054254 De 5/16 a 5/32 in paquete de 10
30053081 De 3/16 a 1/16 in paquete de 10
30053242 De 3/16 a 1/8 in paquete de 10
30053497 De 1/4 a 5/32 in paquete de 10
30053385 De 3/4 a 1/2 in paquete de 10
30054366 De 5/16 a 3/16 in paquete de 10
30054194 De 3/8 a 1/8 in paquete de 10
30053466 De 1/4 a 3/32 in paquete de 10
30054311 De 1/4 a 3/16 in paquete de 10
30052946 De 1 a 5/8 in paquete de 10
30054325 De 1/8 a 3/32 in paquete de 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Código de producto. 30053878 Proveedor Cole-Parmer N.º de proveedor 5010869
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Especificaciones
Certificaciones/conformidad USP;Non-DEHP;Non-Phthalate
Temperatura mín. de funcionamiento (métrico) -35 °C
Cantidad de embalaje 10/Pk.
Tamaño de los tubos De 5/32 a 3/32 in
Puerto de salida Hose Barb Reducer
Método de esterilización E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Forma Straight
Color Natural