Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Código de producto. 30052935 Tienda Cole-Parmer Productos
Change view
Click to view available options
Tamaño de los tubos:
De 1 a 1/2 in
De 1 a 3/4 in
De 1 a 5/8 in
De 1/2 a 1/4 in
De 1/2 a 3/8 in
De 1/2 a 5/16 in.
De 1/4 a 1/16 in
De 1/4 a 1/8 in
De 1/4 a 3/16 in
De 1/4 a 3/32 in
De 1/4 a 5/32 in
De 1/8 a 1/16 in
De 1/8 a 3/32 in
De 3/16 a 1/16 in
De 3/16 a 1/8 in
De 3/16 a 3/32 in
De 3/32 a 1/16 in
De 3/4 a 1/2 in
De 3/4 a 3/8 in
De 3/8 a 1/4 in
De 3/8 a 1/8 in
De 3/8 a 5/16 in
De 3/8 a 5/32 in
De 5/16 a 1/4 in
De 5/16 a 1/8 in
De 5/16 a 3/16 in
De 5/16 a 5/32 in
De 5/32 a 1/8 in
De 5/32 a 3/32 in
De 5/8 a 1/2 in
De 5/8 a 1/4 in
De 5/8 a 3/8 in
De 5/8 a 5/16 in
Tamaño de la unidad:
paquete de 10
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño de los tubos unitSize
30052935 De 1/4 a 3/16 in paquete de 10
30053068 De 3/8 a 5/32 in paquete de 10
30053879 De 5/8 a 5/16 in paquete de 10
30053002 De 3/16 a 3/32 in paquete de 10
30052960 De 1/8 a 1/16 in paquete de 10
30053647 De 1/2 a 3/8 in paquete de 10
30054326 De 5/16 a 1/4 in paquete de 10
30053127 De 5/8 a 1/4 in paquete de 10
30054356 De 1/4 a 5/32 in paquete de 10
30053714 De 3/8 a 1/4 in paquete de 10
30053889 De 5/16 a 3/16 in paquete de 10
30053808 De 5/8 a 3/8 in paquete de 10
30052950 De 1/2 a 5/16 in. paquete de 10
30053607 De 1 a 1/2 in paquete de 10
30053453 De 1/2 a 1/4 in paquete de 10
30054370 De 1/4 a 1/8 in paquete de 10
30053844 De 3/8 a 5/16 in paquete de 10
30054192 De 1/8 a 3/32 in paquete de 10
30053782 De 1 a 3/4 in paquete de 10
30053468 De 3/32 a 1/16 in paquete de 10
30054125 De 5/8 a 1/2 in paquete de 10
30053709 De 5/32 a 3/32 in paquete de 10
30054132 De 1/4 a 3/32 in paquete de 10
30053301 De 1 a 5/8 in paquete de 10
30053431 De 3/4 a 1/2 in paquete de 10
30053733 De 3/4 a 3/8 in paquete de 10
30054112 De 5/16 a 5/32 in paquete de 10
30053870 De 5/16 a 1/8 in paquete de 10
30053146 De 3/16 a 1/16 in paquete de 10
30053484 De 3/8 a 1/8 in paquete de 10
30053023 De 3/16 a 1/8 in paquete de 10
30053569 De 5/32 a 1/8 in paquete de 10
30054262 De 1/4 a 1/16 in paquete de 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Código de producto. 30052935 Proveedor Cole-Parmer N.º de proveedor 5010902
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Temperatura mín. de funcionamiento (métrico) -23°C
Cantidad de embalaje 10/Pk.
Tamaño de los tubos De 1/4 a 3/16 in
Puerto de salida Hose Barb Reducer
Método de esterilización 121°C
Forma Straight
Color Natural