Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Reducing Elbow, White Nylon
Código de producto. p-200105091 Tienda Cole-Parmer Productos
Change view
Click to view available options
Conexión:
1/4 in
1/8 in
Tamaño de la unidad:
paquete de 10
Código de producto. Conexión unitSize
30053293 1/4 in paquete de 10
30053216 1/8 in paquete de 10
2 options
Código de producto. 30053293
Marca: Cole-Parmer 5011746
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifications
Certificaciones/conformidad FDA;Non-DEHP;Non-Phthalate;RoHS
Cantidad de embalaje 10/Pk.
Manguera dentada 1/8 in. Hose Barb
Conexión 1/4 in
Puerto de salida Hose Barb Reducer
Método de esterilización E-beam/gamma irradiation, EtO, or autoclave up to 121°C
Forma Codo
Color Blanco