Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Reducing Elbow, White Nylon
Tienda Cole-Parmer Productos
Click to view available options
Conexión:
1/4 in
1/8 in
Tamaño de la unidad:
paquete de 10
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Especificaciones
Especificaciones
| Certificaciones/conformidad | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Cantidad de embalaje | 10/Pk. |
| Manguera dentada | 1/8 in. Hose Barb |
| Conexión | 1/4 in |
| Puerto de salida | Hose Barb Reducer |
| Método de esterilización | E-beam/gamma irradiation, EtO, or autoclave up to 121°C |
| Forma | Codo |
| Color | Blanco |