Learn More
Cole-Parmer Cleanroom Female Luer Plugs, Straight Adapter
Tienda Cole-Parmer Productos
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nailon
Polipropileno
Color:
Blanco
Natural
Transparente
Conexión:
Conector Luer hembra
Luer hembra con correa
Tamaño de la unidad:
paquete de 10
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;USP |
| Material | Kynar |
| Cantidad de embalaje | 10/Pk. |
| Conexión | Conector Luer hembra |
| Color | Natural |