Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn
Especificaciones
Especificaciones
| Certificaciones/conformidad | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Material | Nailon |
| Cantidad de embalaje | 10/Pk. |
| Conexión | Conector Luer hembra |
| Color | Blanco |