Description
Forms a leak-free luer connection with a quick simple turn. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Specifications
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS |
| Material | Polipropileno |
| Cantidad de embalaje | 10/Pk. |
| Conexión | Female Luer |
| Color | Transparente |