missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clavesin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA535378
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with human and mouse based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2).
Especificaciones
| Clavesin | |
| Polyclonal | |
| Unconjugated | |
| Clvs1 | |
| 4933402J24Rik; A330019N05Rik; bA160A10.4; C6orf212; C6orf213; Cellular retinaldehyde-binding protein-like; clathrin vesicle-associated Sec14 protein 1; clathrin vesicle-associated Sec14 protein 2; clavesin 1; clavesin 2; Clavesin-1; clavesin-1 {ECO:0000303; Clavesin-2; clavesin-2 {ECO:0000303; Clvl1; Clvs1; clvs1 {ECO:0000303; CLVS2; clvs2 {ECO:0000303; CRALBPL; PubMed:19651769}; retinaldehyde binding protein 1-like 1; retinaldehyde binding protein 1-like 2; retinaldehyde-binding protein 1-like 1; Retinaldehyde-binding protein 1-like 2; Retinaldehyde-binding protein 1-like protein 1; retinaldehyde-binding protein 1-like protein 2; RGD1306801; RGD1564200; Rlbp1l1; Rlbp1l2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361459, 366311 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and no preservative | |
| A6JFQ6, A6JUQ6 | |
| Clvs1, Clvs2 | |
| Synthetic peptide corresponding to the C-terminal region of rat CLVS1/2 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido