missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Claudin Antibody Sample Pack

Código de producto. 10333593
Change view
Click to view available options
Cantidad:
80 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10333593 80 μg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10333593

Marca: Novex™ 900900

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Claudin Antibody Sample Pack

The purified, unconjugated antibodies in Claudin Antibody Sample Pack are specific to human, mouse, rat and dog Claudin. Validated applications for antibodies in Claudin Antibody Sample Pack are ELISA, immunofluorescence, tissue staining and Western blotting.

Applications: validated for ELISA, immunofluorescence, tissue staining and Western blotting (Please see the manual for recommended usage for each antibody.)
Reactivity: human, mouse, rat and dog Claudin (Please see the manual for species reactivity of each antibody.)
Product size: 80μg pack size

Tight junctions are specialized regions of cell-cell contact that are particularly abundant in luminal epithelial cell sheets. In freeze-fracture electron micrographs, tight junctions are visualized as belt-like bands of anastomosing sealing strands (TJ strands) that completely encircle the lateral surfaces of each cell. TJ strands on adjacent cells are presumed to interact with each other to form a sort of 'molecular gasket' that prevents ions, water and other molecules from leaking between cells and thus, from one side of the sheet to the other. In addition to this so-called 'barrier' function, the 'fence' function of tight junctions plays an important role in maintaining epithelial cell-polarity by blocking the diffusion of membrane proteins between apical (luminal) and basolateral cell surfaces. Confinement of the glucose symport to apical surfaces allows glucose to be transported vectorially from the lumen, through the cell, and into the bloodstream. Several peripheral membrane proteins are associated with tight junctions including ZO-1, ZO-2, ZO-3 (members of membrane-associated guanylate-kinase family), cingulin, the 7H6 antigen, Rab-3b, symplekin. While their precise functions are not known, roles for these proteins have been suggested in tight junction assembly and maintenance; signal transduction; and the regulation of tight junction permeability.

Furthermore, a growing body of evidence suggests that actin filaments play a major role in regulating tight junction permeability. Until recently, the only transmembrane protein identified in association with tight junctions was occludin, a ∼65 kDa protein with four transmembrane domains. Despite widespread expectation, a critical structural role for occludin in TJ strands was ruled out by the observation of apparently normal tight junctions formed between cells disrupted at both occludin alleles. Fortunately, a closer examination of isolated tight junctions uncovered two related ∼22 kDa, four-transmembrane domain proteins, claudin-1 and claudin-2, with no similarity to occludin. In contrast to occludin, which induces only a small number of short strands at cell-cell contact sites when introduced into fibroblasts lacking tight junctions, claudin-1 and -2 induce networks of strands characteristic of true tight junctions. Though inconclusive, these findings suggest that claudin-1 and -2 are major structural components of TJ strands and that occludin plays some other accessory role. Interest in the tight junction field continues to rise following the discovery of claudins -3, -4, -5, -6, -7, -8, -15, and -16, and experimental results suggesting that tight junctions in different tissues are comprised of distinct sets of claudin family proteins.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Claudin
Aplicaciones Immunofluorescence, Immunohistochemistry, Immunohistochemistry, Western Blot, ELISA
Clasificación Monoclonal/Polyclonal
Clon Monoclonal, polyclonal
Conjugado Unconjugated
Descripción Claudin Antibody Sample Pack, 80μg, Claudin Antigen, Canine, Human, Mouse, Rat Species Reactivity, Affinity Purified, 0.2μm Filtration, Unconjugated, Used for ELISA, Immunofluorescence, Tissue Staining and Western Blotting
génica Claudin-4
Alias de gen CPE-R, CPER, CPETR, CPETR1, WBSCR8, hCPE-R
Especie del huésped Mixed
Método de purificación Affinity Purified
Cantidad 80 μg
Estado normativo RUO
Disciplina de investigación Protein Biology
Primario o secundario Primary
ID de gen (Entrez) 1364
Envase 1 Each
Incluye 0.1% Sodium azide
Especies diana Canine, Human, Mouse, Rat
Contenido y almacenamiento Store in freezer (-5 to -30°C).
Formulario Liquid
Reactividad Canine, Human, Mouse, Rat
Condiciones de envío Wet Ice
Reactividad abrv Cn, Hu, Ms, Rt
Gen diana Claudin
Abrv de aplicación validada IHC, IF, ELISA, WB
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.