Learn More
Claudin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16846084 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16846084 20 μL 20 microlitros
16836084 150 μL 150 microlitros
2 options
Código de producto. 16846084
Marca: Proteintech 164561AP20UL
Rabbit Polyclonal Antibody
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
Especificaciones
Antígeno Claudin 3
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CLDN3
N.º de referencia del gen O15551, Q63400, Q9Z0G9
Alias de gen C7orf1, claudin 3, Claudin3, CLDN3, CPE R 2, CPE R2, CPE receptor 2, CPETR2, HRVP1, RVP1
Símbolos de los genes Cldn3
Especie del huésped Rabbit
Inmunógeno Claudin 3 Fusion Protein Ag9411
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12739, 1365, 65130
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG