Learn More
Claudin 1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13050-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.Especificaciones
| Claudin 1 | |
| Polyclonal | |
| Unconjugated | |
| CLDN1 | |
| claudin 1, CLD1, CLDN1, ILVASC, SEMP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12737, 608207, 9076 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88551, O95832 | |
| CLDN1 | |
| Claudin 1 Fusion Protein Ag3713 | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG |