missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLASP1 Monoclonal Antibody (KT66)

Descripción
The antibody detects a 161 kDa* protein corresponding to the molecular mass of CLASP1 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects CLASP1 by immunocytochemistry in HeLa cells.
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
Especificaciones
Especificaciones
| Antígeno | CLASP1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | KT66 |
| Concentración | ~1.0 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% sodium azide |
| génica | CLASP1 |
| N.º de referencia del gen | Q7Z460, Q80TV8 |
| Alias de gen | 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?