missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKLF Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA543114
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: MDNVQPKIKH RPFCFSVKGH VKMLRLALTV TSMTFFIIAQ APEPYIVITG Sequence homology: Human: 100%.
CKLF is a cytokine. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. CKLF is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. The product of this gene is a cytokine. Cytokines are small proteins that have an essential role in the immune and inflammatory responses. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. The protein encoded by this gene is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones
| CKLF | |
| Polyclonal | |
| Unconjugated | |
| CKLF | |
| 1700001C14Rik; 1810018M11Rik; C32; Chemokine-like factor; chemokine-like factor 1; chemokine-like factor 2; chemokine-like factor 3; chemokine-like factor 4; chemokine-like factor 6; CKLF; CKLF1; Cklf2; CKLF3; CKLF4; CKLF5; Cklf6; HSPC224; transmembrane proteolipid; UCK-1; UNQ410/PRO772 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51192 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UBR5 | |
| CKLF | |
| Synthetic peptide directed towards the N-terminal of human CKLF (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Spot an opportunity for improvement?Share a Content Correction