Learn More
Descripción
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human Jurkat whole cell, human MCF-7 whole cell, human COLO-320 whole cell, human U-87MG whole cell, human A549 whole cell, rat thymus tissue, mouse thymus tissue, mouse testis tissue. ICC/IF: U20S cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Especificaciones
Especificaciones
| Antígeno | cIAP1 |
| Aplicaciones | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 500 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 4mg trehalose and 0.05mg sodium azide |
| génica | BIRC2 |
| N.º de referencia del gen | Q13490, Q62210 |
| Alias de gen | Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW146227; baculoviral IAP repeat containing 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 2; baculoviral IAP repeat-containing protein 2; putative inhibitor of apoptosis; BIRC2; Birc3; cellular inhibitor of apoptosis 1; cIAP1; C-IAP1; cIAP2; HIAP1; HIAP2; Hiap-2; IAP homolog B; IAP1; IAP2; IAP-2; inhibitor of apoptosis protein; inhibitor of apoptosis protein 2; LOC100622859; mcIAP1; MIAP1; MIAP2; mIAP-2; MIHB; MIHC; NFR2-TRAF signalling complex protein; rIAP1; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; RNF48; TNFR2-TRAF-signaling complex protein 2 |
| Símbolos de los genes | BIRC2 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
