missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRNA3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13472367
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13472367 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13472367 Proveedor Invitrogen™ N.º de proveedor PA528258

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4. Predicted reactivity: Mouse (94%), Rat (94%), Chicken (87%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CHRNA3 locus encodes a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CHRNA3
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
génica CHRNA3
N.º de referencia del gen P32297
Alias de gen (a)3; A730007P14Rik; acetylcholine receptor; Acetylcholine receptor alpha 3 (neuronal nicotine); Acra3; Acra-3; alpha 3; cetylcholine receptor alpha 3 (neuronal nicotine); cholinergic receptor; cholinergic receptor nicotinic alpha 3 subunit; cholinergic receptor nicotinic alpha polypeptide 4; cholinergic receptor, nicotinic alpha 3; cholinergic receptor, nicotinic, alpha 3 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 3; CHRNA3; LNCR2; nAChR alpha 3; NACHRA3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha 3 subunit; neuronal nicotinic acetylcholine receptor, alpha3 subunit; PAOD2
Símbolos de los genes CHRNA3
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the Extracellular domain of human Nicotinic Acetylcholine Receptor alpha 3. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1136
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.