missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRM3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13464379
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
13464379 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 13464379 Proveedor Invitrogen™ N.º de proveedor PA532708

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset (100%); Orangutan, Sheep, Bovine, Hamster, Horse (94%); Elephant, Pig (89%); Mouse, Rat, Dog, Rabbit, Opossum, Guinea pig, Turkey, Chicken, Lizard (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CHRM3
Aplicaciones ELISA, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.1% sodium azide
génica CHRM3
N.º de referencia del gen P20309
Alias de gen acetylcholine receptor; acetylcholine receptor, muscarinic 3; AChR M3; cholinergic receptor; cholinergic receptor muscarinic 3; cholinergic receptor, muscarinic 3; cholinergic receptor, muscarinic 3, cardiac; CHRM3; Chrm-3; EGBRS; HM3; M3; m3 muscarinic receptor; M3R; Mm3 mAChR; muscarinic acetylcholine receptor 3; muscarinic acetylcholine receptor M3; PBS
Símbolos de los genes CHRM3
Especie del huésped Rabbit
Inmunógeno Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human CHRM3 / M3.
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1131
Especies diana Human, Monkey
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.