missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHN2 Monoclonal Antibody (E.584.10)

Descripción
It is not recommended to aliquot this antibody.
This gene is a member of the chimerin family and encodes a protein with a phorbol-ester/DAG-type zinc finger, a Rho-GAP domain and an SH2 domain. This protein has GTPase-activating protein activity that is regulated by phospholipid binding and binding of diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane. The protein plays a role in the proliferation and migration of smooth muscle cells. Decreased expression of this gene is associated with high-grade gliomas and breast tumors, and increased expression of this gene is associated with lymphomas. Mutations in this gene have been associated with schizophrenia in men. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Especificaciones
Especificaciones
| Antígeno | CHN2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | E.584.10 |
| Conjugado | Unconjugated |
| Formulación | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| génica | Chn2 |
| N.º de referencia del gen | P52757, Q03070, Q80XD1 |
| Alias de gen | 1700026N20Rik; 4930557O16Rik; ARHGAP3; BCH; beta chimaerin; beta chimerin; beta3-chimaerin; beta-chimaerin; Beta-chimerin; chimerin (chimaerin) 2; chimerin 2; chimerin, beta-2; chimerin, testis-specific; Chn2; CHN2-3; OTTHUMP00000202260; Rho GTPase-activating protein 3; RHOGAP3; rho-GTPase-activating protein 3 |
| Símbolos de los genes | Chn2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?