Learn More
Ácido cloroacético, ACS, 99 %, Thermo Scientific Chemicals
CAS: 79-11-8 | C2H3ClO2 | 94.494 g/mol
40.60€ - 230.15€
CAS 79-11-8
Fórmula molecular C2H3ClO2
Peso molecular (g/mol) 94.494
Número MDL MFCD00002683
Clave InChI FOCAUTSVDIKZOP-UHFFFAOYSA-N
Sinónimo chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
Nombre IUPAC ácido 2-cloroacético
SMILES C(C(=O)O)Cl
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
11348878
SDS Ver documentos
Thermo Scientific Alfa Aesar
041724.22
100 g
40.60€
100 g
11368878
SDS Ver documentos
Thermo Scientific Alfa Aesar
041724.36
500 g
93.97€
500 g
11358878
SDS Ver documentos
Thermo Scientific Alfa Aesar
041724.57
4 x 500 g
230.15€
2000 g
Descripción
Descripción
Chloroacetic acid is used in the production of pharmaceuticals, dyes, detergents, synthetic caffeine, glycine and thioglycolic acid, which acts as a stabilizer in PVC. It acts as a precursor to glyphosate and adrenaline. It also serves as a preservative and bacteriostat. Further, it is used as surface active agents and photosensitive chemicals. It is also used in the O-alkylation of salicylaldehyde and an active component in cosmetics. In addition, it is involved in the preparation of thickening agent carboxymethyl cellulose and carboxymethyl starch.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Mostrar más
Videos
Seguridad y manipulación
Mostrar más
SDS
Documentos
Documentos
Product Certifications
RUO – Research Use Only
Título del producto
Ácido cloroacético, ACS, 99 %, Thermo Scientific Chemicals