Learn More
Invitrogen™ Chlamydia trachomatis MOMP Polyclonal Antibody, HRP

Goat Polyclonal Antibody
Marca: Invitrogen™ PA173074
Descripción
The PA1-73074 antibody reacts with Chlamydia trachomatis MOMP. PA1-73074 has been successfully used in ELISA applications. The PA1-73074 antibody was raised against C. trachomatis serovars (A-K, L1-L3).
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Especificaciones
| Chlamydia trachomatis MOMP | |
| Polyclonal | |
| HRP | |
| C. trachomatis MOMP; Chlamydia MOMP | |
| C. trachomatis serovars (A-K, L1-L3). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal | |
| Goat | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.