missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlamydia trachomatis MOMP Monoclonal Antibody (CL13-227.1.2), Invitrogen™
Descripción
MA1-10670 detects Chlamydia trachomatis MOMP from C. trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, and L3 samples. MA1-10670 has been successfully used in ELISA applications. The MA1-10670 immunogen is C. trachomatis elementary bodies.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
Especificaciones
Especificaciones
| Antígeno | Chlamydia trachomatis MOMP |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | CL13-227.1.2 |
| Concentración | 1.4 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.4 |
| Alias de gen | C. trachomatis MOMP; Chlamydia MOMP |
| Especie del huésped | Mouse |
| Inmunógeno | C. trachomatis elementary bodies. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?