Learn More
DiscoverX ChemiScreen™ SST4 Somatostatin Receptor Membrane Preparation
Description
The ChemiScreen™ SST4 Somatostatin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of SSTR4 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Specifications
| Entrada del ensayo | SST4 |
| Para utilizar con (aplicación) | Membrane Preps |
| Cantidad | 1 vial (200 unidades) |
| Entrada diana | SSTR4 |