Learn More
DiscoverX ChemiScreen™ SST4 Somatostatin Receptor Membrane Preparation
Código de producto. 17305249
Encompass
Change view
Click to view available options
Cantidad:
1 vial (200 unidades)
Tamaño de la unidad:
1 ampolla
Product Code. Cantidad unitSize
17305249 1 vial (200 unidades) 1 ampolla
1 options
This item is not returnable. View return policy
Product Code. 17305249 Supplier DiscoverX Supplier No. HTS125M
This item is not returnable. View return policy
The ChemiScreen™ SST4 Somatostatin Receptor Membrane Preparation allows measurement of ligand binding to SSTR4 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
The ChemiScreen™ SST4 Somatostatin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of SSTR4 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Entrada del ensayo SST4
Para utilizar con (aplicación) Membrane Preps
Cantidad 1 vial (200 unidades)
Entrada diana SSTR4