missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEK2 Mouse anti-Canine, Human, Clone: OTI8D11, liquid, TrueMAB™
Descripción
Checkpoint kinase 2 (Chk2) is a serine/threonine-protein kinase which functions as a cell cycle checkpoint regulator. ATM and ATR kinases are activated in response to ionizing and ultraviolet radition, and phosphorylate Thr68 and other sites in the amino-terminal region of Chk2 dimerisation and activation. Activated Chk2 phosphorylates and inhibits Cdc25C phosphates, preventing the entry into mitosis, and has been shown to stabilize the tumor suppressor protein p53, leading to cell cycle arrest in G1. In addition, Chk2 can phosphorylate BRCA1, allowing BRCA1 to restore survival after DNA damage. Chk2 is a putative tumor suppressor protein, with mutations in to the Chk2 gene linked to Li-Fraumeni syndrome, a familiar cancer usually associated with inherited mutations in TP53. Also, mutations in this gene are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors.
Especificaciones
Especificaciones
| Antígeno | CHEK2 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI8D11 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | CHEK2 |
| N.º de referencia del gen | O96017 |
| Alias de gen | CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?