Learn More
CHCHD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25625-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintece of the MICOS complex stability and the mitochondrial cristae morphology.Especificaciones
| CHCHD3 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD3 | |
| CHCHD3, MIC19, MINOS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54927, 66075 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9CRB9, Q9NX63 | |
| CHCHD3 | |
| CHCHD3 Fusion Protein Ag22579 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |