missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD3 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA519260
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with bovine, canine, ovine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
Especificaciones
| CHCHD3 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD3 | |
| 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 54927 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9NX63 | |
| CHCHD3 | |
| Synthetic peptide sequence (KFKRYESHPVCAD) corresponding to the internal amino acids of CHCHD3 (aa 173-186). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido