missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CES1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | CES1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.56 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.03% ProClin 300; pH 7 |
| génica | CES1 |
| N.º de referencia del gen | P23141 |
| Alias de gen | ACAT; acyl coenzyme A:cholesterol acyltransferase; acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1A; carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 1-like; carboxylesterase 2 (liver); CE-1; CEH; Ces1; Ces-1; Ces1a; Ces1e; Ces1g; Ces1l; CES2; cholesteryl ester hydrolase; cocaine carboxylesterase; Eg; EG244595; Egasyn; Es22; Es-22; ES-HTEL; esterase 22; esterase-22; ES-x; Gm4976; hCE-1; HMSE; HMSE1; human monocyte/macrophage serine esterase 1; liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PCE-1; pI 5.5 esterase; REH; Retinyl ester hydrolase; serine esterase 1; SES1; Ses-1; TGH; triacylglycero; Triacylglycerol hydrolase |
| Símbolos de los genes | CES1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
