Learn More
Ceruloplasmin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21131-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.Especificaciones
| Ceruloplasmin | |
| Polyclonal | |
| Unconjugated | |
| CP | |
| Ceruloplasmin, ceruloplasmin (ferroxidase), CP, CP 2, Ferroxidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12870, 1356 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00450, Q61147 | |
| CP | |
| Ceruloplasmin Fusion Protein Ag15535 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |